Gene Transcripts

Gene Symbol
Pax4
Entrez Gene ID
18506
Gene Name
paired box 4
Gene Aliases
Pax-4
Chromosome Location
Chr. 6: 28442334 - 28449340 on Build GRCm38
UniGene ID
Mm.8026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011038.2 NP_035168.1 9 1000
GenBank mRNA AB008912.1 - 9 975
AB010557.1 - 9 943
AB010558.1 - 9 958
AF031150.1 - 9 1033
AF104231.1 - 8 755
AF145233.1 - 9 947
BC131906.1 - 9 911 40130183
BC131908.1 - 9 909 40130185

View Full Product Details