Gene Transcripts
- Gene Symbol
- Olfr715
- Entrez Gene ID
- 258776
- Gene Name
- olfactory receptor 715
- Gene Aliases
- MOR260-1
- Chromosome Location
- Chr. 7: 107128352 - 107129482 on Build GRCm38
- UniGene ID
- Mm.223207
- Species
- Mus musculus
- Gene Symbol
- Gm10081
- Entrez Gene ID
- 384732
- Gene Name
- predicted gene 10081
- Gene Aliases
- EG384732, ENSMUSG00000060503
- Chromosome Location
- Chr. 7: 107105912 - 107106859 on Build GRCm38
- UniGene ID
- Mm.457863
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC139756.1 |
- |
1 |
925 |
|
View Full Product Details