Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008427.4 | NP_032453.3 | 2 | 434 | |
GenBank mRNA | AB024304.1 | - | 1 | 319 | |
BC120593.1 | - | 2 | 411 | 40129516 | |
BC137594.1 | - | 2 | 411 | 8860613 | |
S71382.1 | - | 2 | 435 | ||
U11075.1 | - | 2 | 404 |