Gene Transcripts

Gene Symbol
Gipc3
Entrez Gene ID
209047
Gene Name
GIPC PDZ domain containing family, member 3
Gene Aliases
Ahl5, Rgs19ip3
Chromosome Location
Chr. 10: 81336540 - 81343744 on Build GRCm38
UniGene ID
Mm.218755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148951.1 NP_683753.1 4 609
GenBank mRNA AB074494.1 - 4 609

View Full Product Details