Gene Transcripts

Gene Symbol
Clec2e
Entrez Gene ID
232409
Gene Name
C-type lectin domain family 2, member e
Gene Aliases
Clra, clr-a
Chromosome Location
Chr. 6: 129091816 - 129100903 on Build GRCm38
UniGene ID
Mm.30870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153506.4 NP_705726.3 5 703
GenBank mRNA BC051091.1 - 5 717

View Full Product Details