Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159389.1 | NP_001152861.1 | 17 | 2012 | |
NM_177306.3 | NP_796280.1 | 14 | 1643 | ||
GenBank mRNA | AK029809.1 | - | 14 | 1644 | |
BC132104.1 | - | 12 | 1326 | 40130381 | |
BC132294.1 | - | 12 | 1326 | 40130571 | |
BC145065.1 | - | 11 | 1208 | ||
FJ804414.1 | - | 16 | 1860 |