Gene Transcripts
- Gene Symbol
- C130073F10Rik
- Entrez Gene ID
- 242574
- Gene Name
- RIKEN cDNA C130073F10 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 101890024 - 101893788 on Build GRCm38
- UniGene ID
- Mm.34251
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA607315.1 |
- |
2 |
228 |
|
AK081744.1 |
- |
3 |
583 |
|
BB374054.2 |
- |
2 |
295 |
|
View Full Product Details