Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301705.1 | NP_001288634.1 | 5 | 742 | |
NM_144899.3 | NP_659148.2 | 5 | 624 | ||
GenBank mRNA | AK155481.1 | - | 5 | 672 | |
AY158701.1 | - | 5 | 565 | ||
BC117925.1 | - | 4 | 537 | 40097608 |