Gene Transcripts

Gene Symbol
Tada2a
Entrez Gene ID
217031
Gene Name
transcriptional adaptor 2A
Gene Aliases
AV319371, D030022J10Rik, Tada2l
Chromosome Location
Chr. 11: 84078920 - 84129642 on Build GRCm38
UniGene ID
Mm.45159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172562.3 NP_766150.1 3 197
GenBank mRNA AK142262.1 - 3 229
AK157938.1 - 3 190
BC038821.1 - 3 200 3990023

View Full Product Details