Gene Transcripts

Gene Symbol
Sbno2
Entrez Gene ID
216161
Gene Name
strawberry notch homolog 2 (Drosophila)
Gene Aliases
BC019206, F730040C21, Sno, Stno
Chromosome Location
Chr. 10: 80056997 - 80106108 on Build GRCm38
UniGene ID
Mm.262102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183426.1 NP_906271.1 18 2117
GenBank mRNA AJ512611.1 - 19 2273
AK089496.1 - 14 2037
AK149806.1 - 18 2141
AK155462.1 - 18 2115
AK155674.1 - 18 2114
BC051234.1 - 1 210
BC056369.1 - 18 2117 6831600
BC064113.1 - 15 1874

View Full Product Details