Gene Transcripts

Gene Symbol
Ms4a3
Entrez Gene ID
170813
Gene Name
membrane-spanning 4-domains, subfamily A, member 3
Gene Aliases
HTm4
Chromosome Location
Chr. 19: 11629499 - 11640851 on Build GRCm38
UniGene ID
Mm.240438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133246.5 NP_573509.1 5 600
GenBank mRNA AF321128.1 - 5 539
AK036464.1 - 5 602
AK158372.1 - 5 601
AY184359.1 - 5 554
BC117013.1 - 5 516 40126332
BC117015.1 - 5 516 40126334
BC144853.1 - 5 516 9053401

View Full Product Details