Gene Transcripts

Gene Symbol
Popdc3
Entrez Gene ID
78977
Gene Name
popeye domain containing 3
Gene Aliases
AA682104, Pop3
Chromosome Location
Chr. 10: 45289305 - 45318450 on Build GRCm38
UniGene ID
Mm.24748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024286.1 NP_077248.1 3, 4 1092
GenBank mRNA AK039691.1 - 3, 4 944
AK052325.1 - 3, 4 1092
BC114993.1 - 3, 4 915 40056724

View Full Product Details