Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006397.2 | NP_006388.2 | 4, 5 | 479 | |
XM_006722619.2 | XP_006722682.1 | 5, 6 | 664 | ||
GenBank mRNA | AK315327.1 | - | 4, 5 | 509 | |
AY363912.1 | - | 4, 5 | 468 | ||
BC011748.2 | - | 4, 5 | 470 | 3160621 | |
BG825340.1 | - | 4, 5 | 483 | ||
CR456841.1 | - | 4, 5 | 419 | ||
Z97029.1 | - | 4, 5 | 479 |