Gene Transcripts

Gene Symbol
Ndufb8
Entrez Gene ID
67264
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8
Gene Aliases
2900010I05Rik, AI987932
Chromosome Location
Chr. 19: 44548576 - 44555446 on Build GRCm38
UniGene ID
Mm.2060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026061.2 NP_080337.1 1, 2 101
GenBank mRNA AK010177.1 - 1, 2 91
AK013516.1 - 1, 2 101
AK028070.1 - 1, 2 112
BC043019.1 - 1, 2 101 5684256

View Full Product Details