Gene Transcripts

Gene Symbol
Tescl
Entrez Gene ID
69301
Gene Name
tescalcin-like
Gene Aliases
1700008P20Rik
Chromosome Location
Chr. 7: 24333080 - 24333965 on Build GRCm38
UniGene ID
Mm.144202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163810.1 NP_001157282.1 1 841
GenBank mRNA AK005787.1 - 1 815
AV266192.1 - 1 208

View Full Product Details