Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039394.1 | NP_001034483.1 | 3 | 1475 | |
NM_133717.3 | NP_598478.1 | 3 | 1181 | ||
GenBank mRNA | AK158283.1 | - | 3 | 1481 | |
AK183544.1 | - | 1 | 151 | ||
AK207605.1 | - | 1 | 151 | ||
BC010248.1 | - | 3 | 1181 | 3155089 |