Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
57764
Gene Name
netrin 4
Gene Aliases
Chromosome Location
Chr. 10: 93641049 - 93745972 on Build GRCm38
UniGene ID
Mm.483868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF268066.1 - 10 1798
AF281278.1 - 10 2051
BC119182.1 - 10 1885 8733931
BC119184.1 - 10 1885 8733933

View Full Product Details