Gene Transcripts

Gene Symbol
Clec7a
Entrez Gene ID
56644
Gene Name
C-type lectin domain family 7, member a
Gene Aliases
BGR, Clecsf12, beta-GR
Chromosome Location
Chr. 6: 129461591 - 129472777 on Build GRCm38
UniGene ID
Mm.239516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020008.2 NP_064392.2 6 778
GenBank mRNA AF262985.1 - 6 774
AY534909.1 - 6 686
BC027742.1 - 6 777 5036647

View Full Product Details