Gene Transcripts

Gene Symbol
Rnasek
Entrez Gene ID
52898
Gene Name
ribonuclease, RNase K
Gene Aliases
2310033H11Rik, AA571397, D11Bwg0434e
Chromosome Location
Chr. 11: 70238123 - 70239852 on Build GRCm38
UniGene ID
Mm.29852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173742.3 NP_776103.1 3 286
GenBank mRNA AK003548.1 - 3 262
AK009592.1 - 3 281
AK018709.1 - 3 235
AK171975.1 - 3 235
BC021602.1 - 3 214 5102170
BC021603.1 - 3 214 5102465
BC024417.1 - 3 244 5065541
CJ181610.1 - 3 286

View Full Product Details