Gene Transcripts

Gene Symbol
Prkch
Entrez Gene ID
81749
Gene Name
protein kinase C, eta
Gene Aliases
Chromosome Location
Chr. 6: 96479243 - 96677368 on Build Rnor_6.0
UniGene ID
Rn.10398
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031085.2 NP_112347.1 1 402
GenBank mRNA BC061763.1 - 1 577
BC081782.1 - 1 402 7123785
X68400.1 - 1 243

View Full Product Details