Gene Transcripts

Gene Symbol
Aifm1
Entrez Gene ID
83533
Gene Name
apoptosis-inducing factor, mitochondrion-associated 1
Gene Aliases
Aif, Pdcd8
Chromosome Location
Chr. X: 135304063 - 135343062 on Build Rnor_6.0
UniGene ID
Rn.203165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031356.1 NP_112646.1 2 199
GenBank mRNA AB041723.1 - 2 215
AF375656.1 - 2 125
AY787821.1 - 2 133
BC072697.1 - 2 157 7105761

View Full Product Details