Gene Transcripts

Gene Symbol
Fgf11
Entrez Gene ID
170632
Gene Name
fibroblast growth factor 11
Gene Aliases
Chromosome Location
Chr. 10: 56403241 - 56409042 on Build Rnor_6.0
UniGene ID
Rn.73083
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130816.1 NP_570829.1 4 429
GenBank mRNA AB079263.1 - 4 429

View Full Product Details