Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139325.3 | NP_647541.1 | 7 | 637 | |
XM_006237330.2 | XP_006237392.1 | 7 | 923 | ||
GenBank mRNA | AF019973.1 | - | 7 | 637 | |
BC060310.1 | - | 7 | 628 | 6888358 | |
FQ214264.1 | - | 7 | 609 | ||
M11931.1 | - | 7 | 637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003750657.3 | XP_003750705.1 | 7 | 693 | |
XM_006244636.2 | XP_006244698.1 | 7 | 925 |