Gene Transcripts

Gene Symbol
Asic3
Entrez Gene ID
286920
Gene Name
acid-sensing (proton-gated) ion channel 3
Gene Aliases
Accn3
Chromosome Location
Chr. 4: 7287868 - 7292917 on Build Rnor_6.0
UniGene ID
Rn.24225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173135.1 NP_775158.1 1 485
XM_008762632.1 XP_008760854.1 1 501
GenBank mRNA AF013598.1 - 1 485

View Full Product Details