Gene Transcripts

Gene Symbol
Sc5d
Entrez Gene ID
114100
Gene Name
sterol-C5-desaturase
Gene Aliases
Sc5dl
Chromosome Location
Chr. 8: 46528066 - 46537014 on Build Rnor_6.0
UniGene ID
Rn.17462
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053642.2 NP_446094.1 5, 6 576
GenBank mRNA AB052846.1 - 4, 5 554
BC081704.1 - 5, 6 576 7131154

View Full Product Details