Gene Transcripts

Gene Symbol
LOC286992
Entrez Gene ID
286992
Gene Name
peptide HP (rs14)
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.127832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173326.1 NP_775448.1 604
GenBank mRNA AF012282.1 - 604

View Full Product Details