Gene Transcripts
- Gene Symbol
- Ncf1
- Entrez Gene ID
- 114553
- Gene Name
- neutrophil cytosolic factor 1
- Gene Aliases
- Ncf-1, p47phox
- Chromosome Location
- Chr. 12: 25497104 - 25506300 on Build Rnor_6.0
- UniGene ID
- Rn.38575
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053734.2 |
NP_446186.2
|
11 |
2513 |
|
GenBank mRNA |
BC061810.1 |
- |
11 |
2513 |
|
View Full Product Details