Gene Transcripts

Gene Symbol
Ralb
Entrez Gene ID
116546
Gene Name
v-ral simian leukemia viral oncogene homolog B
Gene Aliases
dRalb
Chromosome Location
Chr. 13: 35494721 - 35531485 on Build Rnor_6.0
UniGene ID
Rn.4586
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053821.2 NP_446273.1 3 398
GenBank mRNA BC072505.1 - 3 398
L19699.1 - 2 246

View Full Product Details