Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278732.1 | NP_001265661.1 | 2 | 448 | |
NM_012208.3 | NP_036340.1 | 2 | 348 | ||
XM_011537619.1 | XP_011535921.1 | 3 | 361 | ||
XM_011537620.1 | XP_011535922.1 | 3 | 361 | ||
XM_017009287.1 | XP_016864776.1 | 2 | 348 | ||
XM_017009288.1 | XP_016864777.1 | 3 | 412 | ||
XM_017009289.1 | XP_016864778.1 | 3 | 405 | ||
XM_017009290.1 | XP_016864779.1 | 2 | 398 | ||
XM_017009291.1 | XP_016864780.1 | 2 | 91 | ||
XM_017009292.1 | XP_016864781.1 | 2 | 35 | ||
GenBank mRNA | AK293269.1 | - | 2 | 228 | |
AK309832.1 | - | 2 | 446 | ||
AK311277.1 | - | 2 | 444 | ||
AK312976.1 | - | 2 | 166 | ||
BC007680.2 | - | 2 | 167 | 3628006 | |
BC014982.1 | - | 2 | 312 | 4859849 | |
BQ228600.1 | - | 2 | 182 | ||
DA150729.1 | - | 2 | 350 | ||
U18937.1 | - | 2 | 274 |