Gene Transcripts

Gene Symbol
Adk
Entrez Gene ID
25368
Gene Name
adenosine kinase
Gene Aliases
AK
Chromosome Location
Chr. 15: 3033535 - 3435888 on Build Rnor_6.0
UniGene ID
Rn.64165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012895.3 NP_037027.2 8, 9 767
XM_006251631.1 XP_006251693.1 8, 9 881
GenBank mRNA BC081712.1 - 8, 9 767
FQ209450.1 - 3, 4 236
FQ218369.1 - 4, 5 430
FQ219573.1 - 2, 3 75
FQ225093.1 - 4, 5 438
U90340.1 - 8, 9 770

View Full Product Details