Gene Transcripts

Gene Symbol
Hnf1b
Entrez Gene ID
25640
Gene Name
HNF1 homeobox B
Gene Aliases
LF-B3, Tcf2, VHNF1
Chromosome Location
Chr. 10: 71159984 - 71218607 on Build Rnor_6.0
UniGene ID
Rn.11342
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013103.1 NP_037235.1 6, 7 1230
GenBank mRNA BC081826.1 - 5, 6 1345
X56546.1 - 6, 7 1230

View Full Product Details