Gene Transcripts

Gene Symbol
Cd38
Entrez Gene ID
25668
Gene Name
CD38 molecule
Gene Aliases
ADPRC 1
Chromosome Location
Chr. 14: 71714768 - 71754953 on Build Rnor_6.0
UniGene ID
Rn.11414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013127.1 NP_037259.1 6 720
GenBank mRNA D29646.1 - 6 720
D30795.1 - 6 715
FQ212353.1 - 6 725

View Full Product Details