Gene Transcripts

Gene Symbol
Kcnh3
Entrez Gene ID
27150
Gene Name
potassium channel, voltage gated eag related subfamily H, member 3
Gene Aliases
Bec1, Elk2
Chromosome Location
Chr. 7: 140788987 - 140910422 on Build Rnor_6.0
UniGene ID
Rn.144567
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017108.1 NP_058804.1 1, 2 237
GenBank mRNA AB022697.1 - 1, 2 237
AJ007627.1 - 1, 2 102

View Full Product Details