Gene Transcripts

Gene Symbol
Polb
Entrez Gene ID
29240
Gene Name
polymerase (DNA directed), beta
Gene Aliases
Chromosome Location
Chr. 16: 74237001 - 74260272 on Build Rnor_6.0
UniGene ID
Rn.9346
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017141.2 NP_058837.2 5 290
GenBank mRNA BC098668.1 - 5 290
J02776.1 - 5 278
M13961.1 - 5 268
U38801.1 - 285

View Full Product Details