Gene Transcripts
- Gene Symbol
- Ap1b1
- Entrez Gene ID
- 29663
- Gene Name
- adaptor-related protein complex 1, beta 1 subunit
- Gene Aliases
- Adtb1
- Chromosome Location
- Chr. 14: 85250044 - 85281632 on Build Rnor_6.0
- UniGene ID
- Rn.37383
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017277.1 |
NP_058973.1
|
5 |
647 |
|
GenBank mRNA |
M77245.1 |
- |
5 |
647 |
|
View Full Product Details