Gene Transcripts

Gene Symbol
Ptprs
Entrez Gene ID
25529
Gene Name
protein tyrosine phosphatase, receptor type, S
Gene Aliases
Larptp2, Ptprd, Ptpsigma, Ptpsigma., Rptpd
Chromosome Location
Chr. 9: 10585360 - 10646205 on Build Rnor_6.0
UniGene ID
Rn.91202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019140.2 NP_062013.1 3 306
GenBank mRNA BC105753.1 - 549
L12329.1 - 3 306
L19180.1 - 152
L19181.1 - 2 152
L19933.1 - 2 220

View Full Product Details