Gene Transcripts
- Gene Symbol
- Cps1
- Entrez Gene ID
- 497840
- Gene Name
- carbamoyl-phosphate synthetase 1
- Gene Aliases
- cps
- Chromosome Location
- Chr. 9: 74124155 - 74235368 on Build Rnor_6.0
- UniGene ID
- Rn.53968
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017072.1 |
NP_058768.1
|
1 |
86 |
|
GenBank mRNA |
M11710.1 |
- |
1 |
225 |
|
View Full Product Details