Gene Transcripts
- Gene Symbol
- Olr1078
- Entrez Gene ID
- 299593
- Gene Name
- olfactory receptor 1078
- Gene Aliases
- Chromosome Location
- Chr. 7: 13466347 - 13467348 on Build Rnor_6.0
- UniGene ID
- Rn.120045
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207597.1 |
NP_997480.1
|
1 |
940 |
|
GenBank mRNA |
M64376.1 |
- |
1 |
940 |
|
View Full Product Details