Gene Transcripts
- Gene Symbol
- FTCD
- Entrez Gene ID
- 10841
- Gene Name
- formimidoyltransferase cyclodeaminase
- Gene Aliases
- LCHC1
- Chromosome Location
- Chr. 21: 46135981 - 46156481 on Build GRCh38
- UniGene ID
- Hs.415846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006657.2 |
NP_006648.1
|
13, 14 |
1588 |
|
NM_206965.1 |
NP_996848.1
|
13, 14 |
1588 |
|
XM_006723961.3 |
XP_006724024.2
|
13, 14 |
2008 |
|
XM_011529437.2 |
XP_011527739.1
|
13, 14 |
2010 |
|
GenBank mRNA |
AF169017.1 |
- |
13, 14 |
1588 |
|
AF289021.1 |
- |
13, 14 |
1588 |
|
AF289023.1 |
- |
12, 13 |
1544 |
|
AF289024.1 |
- |
13, 14 |
1613 |
|
BC032037.1 |
- |
3, 4 |
875 |
|
BC052248.1 |
- |
13, 14 |
1584 |
6270063 |
BC136383.1 |
- |
13, 14 |
1580 |
9020370 |
BC136395.1 |
- |
13, 14 |
1580 |
9020382 |
BI966164.1 |
- |
4, 5 |
426 |
|
BM021970.1 |
- |
4, 5 |
423 |
|
U91541.1 |
- |
4, 5 |
359 |
|
View Full Product Details