Gene Transcripts

Gene Symbol
RGD1311745
Entrez Gene ID
301079
Gene Name
similar to RIKEN cDNA 1110059G10
Gene Aliases
Chromosome Location
Chr. 8: 132029618 - 132032971 on Build Rnor_6.0
UniGene ID
Rn.161980
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008329.1 NP_001008330.1 3 430
GenBank mRNA BC085913.1 - 3 430
FQ226096.1 - 3 462

View Full Product Details