Gene Transcripts

Gene Symbol
Rnase9
Entrez Gene ID
364301
Gene Name
ribonuclease, RNase A family, 9 (non-active)
Gene Aliases
Chromosome Location
Chr. 15: 27964000 - 27968477 on Build Rnor_6.0
UniGene ID
Rn.128555
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008561.1 NP_001008561.1 2 721
GenBank mRNA AY303815.1 - 2 721

View Full Product Details