Gene Transcripts

Gene Symbol
Smg6
Entrez Gene ID
103677
Gene Name
Smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
AI317223, AU041178
Chromosome Location
Chr. 11: 74925872 - 75164448 on Build GRCm38
UniGene ID
Mm.288460
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002764.1 NP_001002764.1 2 1343
GenBank mRNA BC047279.1 - 2 1354
BC066040.1 - 2 1343 6410245
BC077715.1 - 2 1306

View Full Product Details