Gene Transcripts

Gene Symbol
Hexa
Entrez Gene ID
300757
Gene Name
hexosaminidase A
Gene Aliases
Chromosome Location
Chr. 8: 64325435 - 64350775 on Build Rnor_6.0
UniGene ID
Rn.92939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004443.1 NP_001004443.1 8 929
XM_006243162.1 XP_006243224.1 9 799
GenBank mRNA BC082097.1 - 8 929

View Full Product Details