Gene Transcripts

Gene Symbol
Spata32
Entrez Gene ID
287747
Gene Name
spermatogenesis associated 32
Gene Aliases
RGD1359156, Tex34, Vad1.2
Chromosome Location
Chr. 10: 91291320 - 91302155 on Build Rnor_6.0
UniGene ID
Rn.108312
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005531.1 NP_001005531.1 1, 2 85
GenBank mRNA BC082079.1 - 1, 2 85

View Full Product Details