Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170433.1 | NP_001163904.1 | 8 | 971 | |
NM_026221.2 | NP_080497.1 | 8 | 971 | ||
GenBank mRNA | AK146575.1 | - | 8 | 955 | |
AK154482.1 | - | 8 | 916 | ||
AK173133.1 | - | 8 | 876 | ||
BC035209.1 | - | 8 | 819 | ||
BC072603.1 | - | 8 | 951 | 6830850 |