Gene Transcripts

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
Chromosome Location
Chr. 12: 121308245 - 121354209 on Build GRCh38
UniGene ID
Hs.7101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137559.1 NP_001131031.1 3 215
NM_016237.4 NP_057321.2 3 437
XM_005253900.3 XP_005253957.1 3 439
XM_017019423.1 XP_016874912.1 3 155
XM_017019424.1 XP_016874913.1 3 155
GenBank mRNA AF191339.1 - 3 315
AI879332.1 - 3 439
AK025614.1 - 3 215
AK315451.1 - 3 383
BC001081.2 - 3 370 2823401
BC001950.1 - 3 370 2823401
BC006301.2 - 3 374 4100632
BC034243.1 - 2 77

View Full Product Details