Gene Transcripts

Gene Symbol
SPATA4
Entrez Gene ID
132851
Gene Name
spermatogenesis associated 4
Gene Aliases
SPEF1B, TSARG2
Chromosome Location
Chr. 4: 176184574 - 176195671 on Build GRCh38
UniGene ID
Hs.481235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144644.3 NP_653245.2 1 135
GenBank mRNA AY040204.3 - 1 135
BC021731.2 - 1 36 5269042
HY030172.1 - 1 96

View Full Product Details