Gene Transcripts

Gene Symbol
Arih1
Entrez Gene ID
300756
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Gene Aliases
Chromosome Location
Chr. 8: 64166359 - 64268555 on Build Rnor_6.0
UniGene ID
Rn.29865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013108.3 NP_001013126.2 14 1909
GenBank mRNA AW141789.1 - 4 496
BC083881.1 - 12 1794 7131418

View Full Product Details