Gene Transcripts

Gene Symbol
OR2Y1
Entrez Gene ID
134083
Gene Name
olfactory receptor family 2 subfamily Y member 1
Gene Aliases
OR5-2
Chromosome Location
Chr. 5: 180739123 - 180740058 on Build GRCh38
UniGene ID
Hs.554719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001657.1 NP_001001657.1 1 820
GenBank mRNA BC140756.1 - 1 861

View Full Product Details