Gene Transcripts

Gene Symbol
Cdh20
Entrez Gene ID
363948
Gene Name
cadherin 20
Gene Aliases
Cad20
Chromosome Location
Chr. 13: 24846480 - 24913440 on Build Rnor_6.0
UniGene ID
Rn.62576
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012748.1 NP_001012766.1 3, 4 664
GenBank mRNA AB121033.1 - 3, 4 664

View Full Product Details